missing translation for 'onlineSavingsMsg'
Learn More

Zinc Metal Powder (Dust), Reagents

Catalog No. 01336871
Change view
Click to view available options
Quantity:
500 g
3 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336871 3 kg
01336870 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336871 Supplier Labchem Inc Supplier No. C23505003A5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Zinc Metal Powder (Dust), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
IUPAC Name zinc
SMILES [Zn]

Specifications

Color Silver-Gray
Boiling Point 907°C
Avg. Mol. Wt. or Mol. Wt. Range 65.38
Quantity 3 kg
Packaging Poly Bottle
Form Solid
Chemical Name or Material Zinc Metal Powder (Dust)