missing translation for 'onlineSavingsMsg'
Learn More

Zinc Metal, Mossy, 99.8%, Spectrum™ Chemical

Catalog No. p-7166748
Change view
Click to view available options
Quantity:
12 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614769 12 kg Poly Pail
18614771 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18614769 Supplier Spectrum Chemical Mfg Cor Supplier No. Z102012KG
Only null left

Chemical Identifiers

CAS 7440-66-6
Molecular Weight (g/mol) 65.39

Specifications

CAS Max % 1
Quantity 12 kg
Packaging Poly Pail
Form Granular
Percent Purity 99.8%
Grade Reagent
Chemical Name or Material Zinc Metal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.