missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Metal, 5 x 0.25 x 0.25 in., Sticks, 99.8%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 500 g |
| Packaging | Fiber Tube |
| Particle Size | 5 x 0.25 x 0.25 in. |
| Form | Stick |
| Percent Purity | 99.8% |
| Grade | Reagent |
| Chemical Name or Material | Zinc Metal |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.