missing translation for 'onlineSavingsMsg'
Learn More

Zinc Metal, 5 x 0.25 x 0.25 in., Sticks, 99.8%, Spectrum™ Chemical

Catalog No. 18614761
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Fiber Tube
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614761 500 g Fiber Tube
18614762 2.5 kg Fiber Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18614761 Supplier Spectrum Chemical Mfg Cor Supplier No. Z1040500GM

Chemical Identifiers

CAS 7440-66-6
Molecular Weight (g/mol) 65.39

Specifications

CAS Max % 1
Quantity 500 g
Packaging Fiber Tube
Particle Size 5 x 0.25 x 0.25 in.
Form Stick
Percent Purity 99.8%
Grade Reagent
Chemical Name or Material Zinc Metal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.