missing translation for 'onlineSavingsMsg'
Learn More

Zinc Metal, 325 Mesh, Dust, 95%, Spectrum™ Chemical

Catalog No. 18614760
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614760 500 g Poly Bottle
18614758 2.5 kg Poly Bottle
18614757 12 kg Poly Pail
18614759 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18614760 Supplier Spectrum Chemical Mfg Cor Supplier No. Z1005500GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7440-66-6

Specifications

CAS Max % 1
Quantity 500 g
Mesh Size Approx. 325
Packaging Poly Bottle
Form Dust
Percent Purity 95%
Grade Reagent
Chemical Name or Material Zinc Metal

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.