missing translation for 'onlineSavingsMsg'
Learn More

Zinc Metal, 325 Mesh, Dust, 95%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor Z100525KG

 View more versions of this product

Catalog No. 18614758


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Zinc Metal
1
Approx. 325
Dust
Reagent
7440-66-6
2.5 kg
Poly Bottle
95%

Chemical Identifiers

7440-66-6
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.