missing translation for 'onlineSavingsMsg'
Learn More

Zinc Metal, 20 Mesh, Granular, Spectrum™ Chemical

Catalog No. 18614755
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Fiber Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614755 500 g Fiber Drum
18614752 100 lb. Fiber Drum
18614753 2.5 kg Fiber Drum
18614754 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18614755 Supplier Spectrum Chemical Mfg Cor Supplier No. Z1019500GM
Only null left

Chemical Identifiers

CAS 7440-66-6
Molecular Weight (g/mol) 65.39

Specifications

CAS Max % 1
Quantity 500 g
Packaging Fiber Drum
Particle Size Approx. 20 Mesh
Form Granular
Grade Ungraded
Chemical Name or Material Zinc Metal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.