missing translation for 'onlineSavingsMsg'
Learn More

Zinc Metal, 0.62 mm thick, Sheet, 99.9%, Spectrum™ Chemical

Catalog No. 18614751
Change view
Click to view available options
Quantity:
500 g
Packaging:
Fiber Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614751 500 g Fiber Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18614751 Supplier Spectrum Chemical Mfg Cor Supplier No. Z1033500GM

Chemical Identifiers

CAS 7440-66-6
Molecular Weight (g/mol) 65.39

Specifications

CAS Max % 1
Quantity 500 g
Packaging Fiber Tube
Form Sheet
Percent Purity 99.9%
Grade Ungraded
Chemical Name or Material Zinc Metal

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.