missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Metal, 0.25 in. and Finer, Shot, 99.8%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 500 g |
| Packaging | Poly Bottle |
| Particle Size | ≤0.25 in. |
| Form | Shot |
| Percent Purity | 99.8% |
| Grade | Reagent |
| Chemical Name or Material | Zinc Metal |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.