missing translation for 'onlineSavingsMsg'
Learn More

Zinc meso-tetraphenylporphine

Catalog No. p-7050822
Change view
Click to view available options
Quantity:
0.5 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3955202 0.5 g
AA3955204 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3955202 Supplier Thermo Scientific Chemicals Supplier No. 03955202
Only null left

CAS: 14074-80-7 | C44H28N4Zn | 678.12 g/mol

A porphyrin used in light emitting diodes. It has valuable biological applications in molecular biology, fluoroimmunoassay and new pharmaceutical developments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14074-80-7
Molecular Formula C44H28N4Zn
Molecular Weight (g/mol) 678.12
MDL Number MFCD00012155,MFCD00012155
InChI Key FGEZGTKFMCNEOZ-UHFFFAOYSA-N
Synonym zinc meso-tetraphenylporphine
PubChem CID 91667917
IUPAC Name 5,10,15,20-tetraphenylporphyrin-22,24-diide;zinc
SMILES [Zn].[N-]1C2=CC=C1C(=C1C=CC(=N1)C(=C1[N-]C(C=C1)=C(C1=NC(C=C1)=C2C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Purple
Odor Odorless
Quantity 0.5 g
Solubility Information Insoluble in water.
Formula Weight 678.1
Physical Form Crystalline
Chemical Name or Material Zinc meso-tetraphenylporphine
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only