missing translation for 'onlineSavingsMsg'
Learn More

Zinc iron oxide

Catalog No. p-7049723
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1940714 25 g
AA1940722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1940714 Supplier Thermo Scientific Chemicals Supplier No. 01940714

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12063-19-3 | Fe2H2O4Zn | 243.082 g/mol

Zinc iron oxide is used as pigments, especially in applications related to heat stability. It is also used in high corrosion-resistant coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12063-19-3
Molecular Formula Fe2H2O4Zn
Molecular Weight (g/mol) 243.082
MDL Number MFCD00049621
InChI Key NNGHIEIYUJKFQS-UHFFFAOYSA-L
Synonym Zinc ferrite
PubChem CID 11831558
IUPAC Name hydroxy(oxo)iron;zinc
SMILES O[Fe]=O.O[Fe]=O.[Zn]

Specifications

Physical Form Fine Powder
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 241.06
Odor Odorless
Chemical Name or Material Zinc iron oxide
EINECSNumber 235-052-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only