missing translation for 'onlineSavingsMsg'
Learn More

Zinc iodide, 99.999%, (trace metal basis), extra pure

CAS: 10139-47-6 | I2Zn | 319.19 g/mol

$129.56 - $129.56

Chemical Identifiers

CAS 10139-47-6
Molecular Formula I2Zn
Molecular Weight (g/mol) 319.19
MDL Number MFCD00011299
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC212760050
SDS View Documents
Thermo Scientific Chemicals
212760050
5 g Glass Bottle
Each for $129.56
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Chemical Identifiers

10139-47-6
I2Zn
319.19
MFCD00011299

Specifications

446°C
5 g
I2Zn
MFCD00011299
15, 10339
319.19
100.00%
Glass Bottle
4.7400g/mL
10139-47-6
99.999%
ZnI2
01,1294; 05,765; 10,462; 11,604; 12,576; 13,88
Solubility in water: 4500g/L (20°C). Other solubilities: soluble in alkalies,soluble in alcohol and ether
319.19
Trace Metal Basis
4.74
Zinc iodide, 99.999%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 233-396-0

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only