missing translation for 'onlineSavingsMsg'
Learn More

Zinc iodide, 98%

Catalog No. AA1166118
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1166118 50 g
AA1166130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1166118 Supplier Thermo Scientific Chemicals Supplier No. 01166118

CAS: 10139-47-6

Zinc iodide is often used as an x-ray opaque penetrant in industrial radiography to improve the contrast between the damage and intact composite.n conjunction with osmium tetroxide ZnI2 is used as a stain in electron microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10139-47-6
MDL Number MFCD00011299

Specifications

Boiling Point 625°C (decomposition)
Melting Point 446°C
Color White to Yellow
Physical Form Crystalline
Quantity 50 g
Linear Formula ZnI2
UN Number UN3260
Merck Index 14,10140
Solubility Information Freely soluble in water, glycerol, alcohol, ether
Formula Weight 319.18
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 4.74 g/mL
Chemical Name or Material Zinc iodide
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 233-396-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only