missing translation for 'onlineSavingsMsg'
Learn More

Zinc(II) Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. p-7136703
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12945G 5 g
T129425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T12945G Supplier TCI America Supplier No. T12945G
Only null left

Chemical Identifiers

CAS 54010-75-2
Molecular Formula C2F6O6S2Zn
Molecular Weight (g/mol) 363.51
MDL Number MFCD00013229
InChI Key CITILBVTAYEWKR-UHFFFAOYSA-L
Synonym zinc trifluoromethanesulfonate, zinc ii trifluoromethanesulfonate, zinc trifluoromethanesulphonate, zinc triflate, zinc ii triflate, zn otf 2, methanesulfonic acid, trifluoro-, zinc salt, zinc 2+ ion ditriflate, methanesulfonic acid, 1,1,1-trifluoro-, zinc salt 2:1, zinc ii trifluoromethanesulphonate
PubChem CID 104671
IUPAC Name zinc(2+) ditrifluoromethanesulfonate
SMILES [Zn++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Color White
Physical Form Crystalline Powder
Quantity 5 g
UN Number 1759
Formula Weight 363.51
Percent Purity ≥98.0% (T)
Chemical Name or Material Zinc(II) Trifluoromethanesulfonate