missing translation for 'onlineSavingsMsg'
Learn More

Zinc(II) Tetraphenylporphyrin 98.0+%, TCI America™

Catalog No. p-7138006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z00361G 1 g
Z00365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Z00361G Supplier TCI America Supplier No. Z00361G
Only null left

Chemical Identifiers

CAS 14074-80-7
Molecular Formula C44H28N4Zn
Molecular Weight (g/mol) 678.12
MDL Number MFCD00012155,MFCD00012155
InChI Key FGEZGTKFMCNEOZ-UHFFFAOYSA-N
Synonym zinc meso-tetraphenylporphine
PubChem CID 91667917
IUPAC Name 2,7,12,17-tetraphenyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(20),2,4,6(24),7,9,11,13(22),14,16,18-undecaene-21,23-diide zinc
SMILES [Zn].[N-]1C2=CC=C1C(=C1C=CC(=N1)C(=C1[N-]C(C=C1)=C(C1=NC(C=C1)=C2C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Purple
Quantity 1 g
Formula Weight 678.12
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Zinc(II) Tetraphenylporphyrin