missing translation for 'onlineSavingsMsg'
Learn More

Zinc(II) Gluconate Hydrate 98.0+%, TCI America™

Catalog No. G027725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G027725G 25 g
G0277500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G027725G Supplier TCI America Supplier No. G027725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4468-02-4
Molecular Formula C12H22O14Zn
Molecular Weight (g/mol) 455.67
MDL Number MFCD00868110
InChI Key WHMDKBIGKVEYHS-IYEMJOQQSA-L
Synonym zinc ii gluconate hydrate, zinc 2+ bis d-gluconate hydrate
PubChem CID 131674739
IUPAC Name zinc(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
SMILES [Zn++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

Specifications

Color White
Quantity 25 g
Formula Weight 455.67
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Zinc(II) Gluconate Hydrate
EINECSNumber (2)-3129
RTECSNumber ZH3750000
TSCA Yes