missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc(II) Dibutyldithiocarbamate 98.0+%, TCI America™
$72.70 - $352.30
Chemical Identifiers
| CAS | 136-23-2 |
|---|---|
| Molecular Formula | C18H36N2S4Zn |
| Molecular Weight (g/mol) | 474.12 |
| MDL Number | MFCD00067274 |
| InChI Key | BOXSVZNGTQTENJ-UHFFFAOYSA-L |
| Synonym | Dibutyldithiocarbamic Acid Zinc(II) Salt |
| PubChem CID | 5284483 |
| IUPAC Name | zinc;N,N-dibutylcarbamodithioate |
| SMILES | CCCCN(CCCC)C(=S)[S-].CCCCN(CCCC)C(=S)[S-].[Zn+2] |
Chemical Identifiers
| 136-23-2 | |
| 474.12 | |
| BOXSVZNGTQTENJ-UHFFFAOYSA-L | |
| 5284483 | |
| CCCCN(CCCC)C(=S)[S-].CCCCN(CCCC)C(=S)[S-].[Zn+2] |
| C18H36N2S4Zn | |
| MFCD00067274 | |
| Dibutyldithiocarbamic Acid Zinc(II) Salt | |
| zinc;N,N-dibutylcarbamodithioate |
Specifications
| 136-23-2 | |
| White-Yellow | |
| MFCD00067274 | |
| 3077 | |
| BOXSVZNGTQTENJ-UHFFFAOYSA-L | |
| zinc;N,N-dibutylcarbamodithioate | |
| 5284483 | |
| ≥98.0% (T) | |
| Zinc(II) Dibutyldithiocarbamate |
| 109°C | |
| C18H36N2S4Zn | |
| 25 g | |
| Dibutyldithiocarbamic Acid Zinc(II) Salt | |
| CCCCN(CCCC)C(=S)[S-].CCCCN(CCCC)C(=S)[S-].[Zn+2] | |
| 474.12 | |
| 474.12 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-2122
RTECSNumber : ZH0175000
TSCA : Yes