missing translation for 'onlineSavingsMsg'
Learn More

Zinc(II) Dibutyldithiocarbamate 98.0+%, TCI America™

Catalog No. D0227500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0227500G 500 g
D022725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0227500G Supplier TCI America Supplier No. D0227500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 136-23-2
Molecular Formula C18H36N2S4Zn
Molecular Weight (g/mol) 474.12
MDL Number MFCD00067274
InChI Key BOXSVZNGTQTENJ-UHFFFAOYSA-L
Synonym Dibutyldithiocarbamic Acid Zinc(II) Salt
PubChem CID 5284483
IUPAC Name zinc;N,N-dibutylcarbamodithioate
SMILES CCCCN(CCCC)C(=S)[S-].CCCCN(CCCC)C(=S)[S-].[Zn+2]

Specifications

Melting Point 109°C
Color White-Yellow
Quantity 500 g
UN Number 3077
Formula Weight 474.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Zinc(II) Dibutyldithiocarbamate
EINECSNumber (2)-2122
RTECSNumber ZH0175000
TSCA Yes