missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc(II) Dibutyldithiocarbamate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 109°C |
| Color | White-Yellow |
| Quantity | 25 g |
| UN Number | 3077 |
| Formula Weight | 474.12 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Zinc(II) Dibutyldithiocarbamate |
Safety and Handling
| EINECSNumber | (2)-2122 |
| RTECSNumber | ZH0175000 |
| TSCA | Yes |