missing translation for 'onlineSavingsMsg'
Learn More

Zinc(II) Dibenzyldithiocarbamate 97.0+%, TCI America™

Catalog No. D015725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D015725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D015725G Supplier TCI America Supplier No. D015725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14726-36-4
Molecular Formula C30H28N2S4Zn
Molecular Weight (g/mol) 610.188
MDL Number MFCD00014470
InChI Key AUMBZPPBWALQRO-UHFFFAOYSA-L
Synonym zinc ii dibenzyldithiocarbamate, zinc dibenzyldithiocarbamate, dibenzyldithiocarbamic acid zinc salt, dibenzyldithiocarbamic acid, zinc salt, zinc n,n-dibenzylcarbamodithioate, zinc dibenzyl dithiocarbamate, zinc bis dibenzyldithiocarbamate, acmc-209cyb, ksc910q0r, bis dibenzyldithiocarbamic acid zinc
PubChem CID 84610
IUPAC Name zinc;N,N-dibenzylcarbamodithioate
SMILES C1=CC=C(C=C1)CN(CC2=CC=CC=C2)C(=S)[S-].C1=CC=C(C=C1)CN(CC2=CC=CC=C2)C(=S)[S-].[Zn+2]

Specifications

Melting Point 182°C
Color White
Quantity 25 g
Formula Weight 610.19
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Zinc(II) Dibenzyldithiocarbamate
EINECSNumber (3)-2185
TSCA Yes