missing translation for 'onlineSavingsMsg'
Learn More

Zinc(II) Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. Z00271G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z00271G 1 g
Z00275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Z00271G Supplier TCI America Supplier No. Z00271G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 168106-25-0
Molecular Formula C4F12N2O8S4Zn
Molecular Weight (g/mol) 625.651
MDL Number MFCD16621474
InChI Key QEORIOGPVTWFMH-UHFFFAOYSA-N
Synonym Bis(trifluoromethanesulfonyl)imide Zinc(II) Salt, Zinc(II) Triflimide
PubChem CID 131849436
IUPAC Name zinc;bis(trifluoromethylsulfonyl)azanide
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Zn+2]

Specifications

Melting Point 147°C
Color White
Quantity 1 g
UN Number 1759
Formula Weight 625.65
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Zinc(II) Bis(trifluoromethanesulfonyl)imide