missing translation for 'onlineSavingsMsg'
Learn More

Zinc, Granular, 30 Mesh, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. p-5391685
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02004599 500 g Poly Bottle
02004598 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02004599 Supplier Avantor Supplier No. 424801

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Quantity 500 g
Mesh Size 30 Mesh
Packaging Poly Bottle
Form Granular
Formula Weight 65.39
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Zinc