missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc, granular, 20 mesh
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Specifications
| Color | Gray |
| Melting Point | 420.0°C |
| Boiling Point | 907.0°C |
| Density | 7.1400g/mL |
| Quantity | 2.5 kg |
| Packaging | Plastic bottle |
| Form | Granular Powder |
| Linear Formula | Zn |
| Fieser | 01,1276; 02,459; 03,334; 04,574; 05,753; 06,672; 07,426; 08,532; 10,459; 11,98; 12,568; 13,298; 14,111; 16,106; 17,406 |
| Formula Weight | 65.38 |
| Show More |
RUO – Research Use Only