missing translation for 'onlineSavingsMsg'
Learn More

Zinc Gluconate, USP, 97-102%, Spectrum™ Chemical

Catalog No. 18607850
Change view
Click to view available options
Quantity:
500 g
50 kg
Packaging:
Fiber Drum
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607850 500 g Poly Bottle
18607853 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18607850 Supplier Spectrum Chemical Mfg Cor Supplier No. Z1077500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Z1077, 4468-02-4, C12H22O14Zn

Spectrum™ Chemical Zinc Gluconate, USP is used for treatment and prevention of zinc deficiency. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 4468-02-4
Molecular Formula C12H22O14Zn
Molecular Weight (g/mol) 455.67
MDL Number MFCD00868110
InChI Key WHMDKBIGKVEYHS-IYEMJOQQSA-L
IUPAC Name zinc(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
SMILES [Zn++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

Specifications

CAS Max % 1
pH 5.5 to 7.5
Packaging Poly Bottle
Quantity 500 g
Percent Purity 97 to 102%
Grade USP