missing translation for 'onlineSavingsMsg'
Learn More

Zinc gluconate hydrate, 97%

CAS: 4468-02-4 | C4H7NO | 85.11 g/mol

$104.86 - $284.22

Chemical Identifiers

CAS 4468-02-4
MDL Number MFCD00868110
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2302222
View Documents
Thermo Scientific Chemicals
B2302222
100 g
Each for $104.86
Only null left
 
AAB2302236
View Documents
Thermo Scientific Chemicals
B2302236
500 g
Each for $284.22
Only null left
 
Description

Description

Zinc gluconate hydrate is used as a food additive, a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc gluconate hydrate is used as a food additive, a pharmaceutical intermediate.

Solubility
Soluble in water 100g/ l.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

4468-02-4
MFCD00868110

Specifications

4468-02-4
Odorless
MFCD00868110
14,4456
Soluble in water 100g/ l.
[Zn++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
455.67
455.69 (Anhydrous)
Zinc gluconate hydrate
6.5
C12H22O14Zn
100 g
zinc ii gluconate hydrate, zinc 2+ bis d-gluconate hydrate
WHMDKBIGKVEYHS-IYEMJOQQSA-L
zinc;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate;hydrate
131674739
97%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 224-736-9

RTECSNumber : ZH3750000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only