Learn More
Zinc foil, 1.0mm (0.04in) thick, Puratronic™, 99.9985% (metals basis)
Form: Approx.4.5g/25 x 25mm | CAS: 7440-66-6 | Zn | 65.38 g/mol
Supplier: Thermo Scientific Chemicals 011915FI
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Gray | |
| 7440-66-6 | |
| 50 x 50 mm | |
| 99.9985% (Metals basis) | |
| Zn | |
| dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust | |
| [Zn] | |
| 65.38 | |
| CHEBI:30185 |
| Zinc | |
| Gray foil | |
| Odorless | |
| Foil | |
| MFCD00011291 | |
| HCHKCACWOHOZIP-UHFFFAOYSA-N | |
| zinc | |
| 23994 |
Chemical Identifiers
| 7440-66-6 | |
| 65.38 | |
| HCHKCACWOHOZIP-UHFFFAOYSA-N | |
| 23994 | |
| zinc |
| Zn | |
| MFCD00011291 | |
| dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust | |
| CHEBI:30185 | |
| [Zn] |
Safety and Handling
EINECSNumber : 231-175-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only