missing translation for 'onlineSavingsMsg'
Learn More

Zinc foil, 0.1mm (0.004in) thick, Puratronic™, 99.994% (metals basis)

Catalog No. p-4412872
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 x 100 mm
100 x 300 mm
50 x 50 mm
Catalog No. Quantity
AA11912FI 50 x 50 mm
AA11912GH 100 x 100 mm
AA11912HG 100 x 300 mm
3 options

Catalog No. AA11912FI

Supplier: Thermo Scientific Chemicals 011912FI

Only null left

Form: approx. 4.5g/50 x 50mm | CAS: 7440-66-6 | Zn | 65.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Color Gray
Appearance Gray foil
Density 7.14g/cm3
Quantity 50 x 50 mm
Odor Odorless
Form Foil
Assay Percent Range 99.994% (Metals basis)
Chemical Name or Material Zinc
EINECSNumber 231-175-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only