missing translation for 'onlineSavingsMsg'
Learn More

Zinc fluoride, Puratronic™, 99.995% (metals basis)

CAS: 7783-49-5 | F2Zn | 103.377 g/mol

$155.70 - $1391.05

Chemical Identifiers

CAS 7783-49-5
Molecular Formula F2Zn
Molecular Weight (g/mol) 103.377
MDL Number MFCD00011298
InChI Key BHHYHSUAOQUXJK-UHFFFAOYSA-L
Synonym zinc ii fluoride, zinc 2+ ion difluoride, acmc-1bhy1, ksc379c0l, zinc fluoride 50g, zinc, ion zn2+ difluoride
PubChem CID 10129912
IUPAC Name zinc;difluoride
SMILES [F-].[F-].[Zn+2]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1298606
View Documents
Thermo Scientific Chemicals
01298606
5 g
Each for $155.70
Only null left
 
AA1298614
View Documents
Thermo Scientific Chemicals
01298614
25 g
Each for $488.99
Only null left
 
AA1298622
View Documents
Thermo Scientific Chemicals
01298622
100 g
Each for $1,391.05
Only null left
 
Description

Description

It is widely used in fluorination of organic compounds, as additive of electrolytic aluminum and also in electroplating baths. It is used in preserving food, manufacture of flux, oxygen-sensitive applications such as metal production, glasses, enamels, fabrics and plastics. It is also used in oil refining, manufacture of pharmaceuticals, manufacture phosphors for fluorescent electric lights, and floor coverings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7783-49-5
103.377
BHHYHSUAOQUXJK-UHFFFAOYSA-L
10129912
[F-].[F-].[Zn+2]
F2Zn
MFCD00011298
zinc ii fluoride, zinc 2+ ion difluoride, acmc-1bhy1, ksc379c0l, zinc fluoride 50g, zinc, ion zn2+ difluoride
zinc;difluoride

Specifications

1500°C
7783-49-5
Powder
F2Zn
MFCD00011298
zinc ii fluoride, zinc 2+ ion difluoride, acmc-1bhy1, ksc379c0l, zinc fluoride 50g, zinc, ion zn2+ difluoride
BHHYHSUAOQUXJK-UHFFFAOYSA-L
zinc;difluoride
10129912
99.995%
4.95 g/mL
(metals basis)
872°C
White
5 g
ZnF2
14,10136
Sparingly soluble in hydrochloric acid,nitric acid,and ammonium hydroxide.
[F-].[F-].[Zn+2]
103.377
103.37
Puratronic
Zinc fluoride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c

H315-H318-H335

EINECSNumber : 232-001-9

RTECSNumber : ZH3500000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only