missing translation for 'onlineSavingsMsg'
Learn More

Zinc fluoride, anhydrous, 99% min

CAS: 7783-49-5 | F2Zn | 103.377 g/mol

Supplier:  Thermo Scientific Chemicals 01154130

 View more versions of this product

Catalog No. AA1154130


Only null left
Explore available promotions

Description

Description

It is widely used in fluorination of organic compounds, as additive of electrolytic aluminum and also in electroplating baths. It is used in preserving food, manufacture of flux, oxygen-sensitive applications such as metal production, glasses, enamels, fabrics and plastics. It is also used in oil refining, manufacture of pharmaceuticals, manufacture phosphors for fluorescent electric lights, and floor coverings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1500°C
872°C
White
250 g
F2Zn
zinc ii fluoride, zinc 2+ ion difluoride, acmc-1bhy1, ksc379c0l, zinc fluoride 50g, zinc, ion zn2+ difluoride
BHHYHSUAOQUXJK-UHFFFAOYSA-L
zinc;difluoride
10129912
Odorless
Zinc fluoride
7783-49-5
Powder
99% min. (anhydrous)
MFCD00011298
Sparingly soluble in hydrochloric acid,nitric acid,and ammonium hydroxide.
[F-].[F-].[Zn+2]
103.377
103.37
4.95

Chemical Identifiers

7783-49-5
103.377
BHHYHSUAOQUXJK-UHFFFAOYSA-L
10129912
[F-].[F-].[Zn+2]
F2Zn
MFCD00011298
zinc ii fluoride, zinc 2+ ion difluoride, acmc-1bhy1, ksc379c0l, zinc fluoride 50g, zinc, ion zn2+ difluoride
zinc;difluoride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c

H315-H318-H335

EINECSNumber : 232-001-9

RTECSNumber : ZH3500000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only