missing translation for 'onlineSavingsMsg'
Learn More

Zinc, Dust Particle Size <63μm, MilliporeSigma™

Catalog No. M1087741000
Change view
Click to view available options
Quantity:
1 kg
50 kg
Packaging:
Plastic bottle
Steel drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1087741000 1 kg Plastic bottle
M1087749050 50 kg Steel drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1087741000 Supplier Technidata Supplier No. 1.08774.1000
Only null left

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Appearance Gray solid
Quantity 1 kg
Odor Odorless
Packaging Plastic bottle
Vapor Pressure 1.33hPa
Form Solid
UN Number UN 3077
Formula Weight 65.39
Chemical Name or Material Zinc
Hazard Category H410: Very toxic to aquatic life with long lasting effects.
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 231-175-3