missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc diethyldithiocarbamate, Zn 17-19.5%
Description
Zinc diethyldithiocarbamate is a chelating ligand. It is used as a common accelerator to latex. A fast curing primary or secondary effective ultra-accelerator for natural and synthetic latex compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 178°C to 183°C |
| Color | White |
| Density | 1.48 g/mL |
| pH | 7.8 |
| Flash Point | 204°C (400°F) |
| Odor | Odorless |
| Linear Formula | Zn[CS2N(CH2CH3)2]2 |
| Quantity | 500 g |
| UN Number | UN3077 |
| Solubility Information | Insoluble in water. Soluble in benzene carbon disulfide and organic liquids. |
| Show More |
Safety and Handling
| Hazard Category | H302-H315-H317-H319-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H317-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 238-270-9 |
| RTECSNumber | ZH0350000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only