missing translation for 'onlineSavingsMsg'
Learn More

Zinc Diethyldithiocarbamate 99.0+%, TCI America™

Catalog No. D049225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D049225G 25 g
D0492500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D049225G Supplier TCI America Supplier No. D049225G

Chemical Identifiers

CAS 14324-55-1
Molecular Formula C10H20N2S4Zn
Molecular Weight (g/mol) 361.904
MDL Number MFCD00064798
InChI Key RKQOSDAEEGPRER-UHFFFAOYSA-L
Synonym zinc diethyldithiocarbamate, ethyl ziram, zinc bis diethyldithiocarbamate, diethyldithiocarbamic acid zinc salt, zinc n,n-diethylcarbamodithioate, ethyl zimate, unii-icw4708z8g, ditiocarb zinc, zdc, zdec
PubChem CID 26633
IUPAC Name zinc;N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)[S-].CCN(CC)C(=S)[S-].[Zn+2]

Specifications

Melting Point 181°C
Color White-Yellow
Quantity 25 g
UN Number 3077
Formula Weight 361.90
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Zinc Diethyldithiocarbamate
EINECSNumber (2)-1843
RTECSNumber ZH0350000
TSCA Yes