missing translation for 'onlineSavingsMsg'
Learn More

Zinc copper couple

Catalog No. p-41012
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC271440250 25 g Glass bottle
AC271441000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC271440250 Supplier Thermo Scientific Chemicals Supplier No. 271440250
Only null left

CAS: 53801-63-1 | CuZn | 128.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53801-63-1
Molecular Formula CuZn
Molecular Weight (g/mol) 128.93
MDL Number MFCD00084851
InChI Key TVZPLCNGKSPOJA-UHFFFAOYSA-N
Synonym zinc-copper couple, cu.zn, zinc copper, copper, compound with zinc 1:1, zinc-copper, brass alloy, zn copper, copper-zinc alloy, zinc copper couple, cu-zn
PubChem CID 10290809
IUPAC Name copper;zinc
SMILES [Cu].[Zn]

Specifications

Color Gray to Black
Quantity 25 g
Packaging Glass bottle
Form Powder
Linear Formula ZnCu
Fieser 14,309; 15,367; 16,331; 17,407
Chemical Name or Material Zinc copper couple
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable solid.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep container tightly closed.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only