missing translation for 'onlineSavingsMsg'
Learn More

Zinc-copper couple, Copper content typically ca 1-4%

Catalog No. AAL0981114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0981114 25 g
AAL0981122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0981114 Supplier Thermo Scientific Chemicals Supplier No. L0981114

CAS: 53801-63-1 | CuZn | 128.93 g/mol

Zinc-copper couple is used in the Simmons-Smith cyclopropanation reaction, Reformatsky reaction. It is used as a chemical for the selective, stereospecific reduction of alkynes to cis-alkenes. It is used in for deiodination of iodotributylstannylalkanes. In the presence of a free-radical initiator, it promotes the formylation of perfluoroalkyl iodides with DMF to give fluorinated aldehydes in high yield. It is used in dermatology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc-copper couple is used in the Simmons-Smith cyclopropanation reaction, Reformatsky reaction. It is used as a chemical for the selective, stereospecific reduction of alkynes to cis-alkenes. It is used in for deiodination of iodotributylstannylalkanes. In the presence of a free-radical initiator, it promotes the formylation of perfluoroalkyl iodides with DMF to give fluorinated aldehydes in high yield. It is used in dermatology.

Solubility
Insoluble in water.

Notes
Store at 4°C. Moisture Sensitive. Do not store together with acids. Protect from humidity and water. Incompatible materials are amines, cadmium, chlorinated solvents, Sulfur compounds.

Chemical Identifiers

CAS 53801-63-1
Molecular Formula CuZn
Molecular Weight (g/mol) 128.93
MDL Number MFCD00084851
InChI Key TVZPLCNGKSPOJA-UHFFFAOYSA-N
Synonym zinc-copper couple, cu.zn, zinc copper, copper, compound with zinc 1:1, zinc-copper, brass alloy, zn copper, copper-zinc alloy, zinc copper couple, cu-zn
PubChem CID 10290809
IUPAC Name copper zinc
SMILES [Cu].[Zn]

Specifications

Melting Point 419°C to 420°C
Quantity 25 g
Sensitivity Moisture sensitive
UN Number UN3089
Solubility Information Insoluble in water.
Assay Copper content typically ≈1-3%
Chemical Name or Material Zinc-copper couple
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
EINECSNumber 684-244-9
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only