missing translation for 'onlineSavingsMsg'
Learn More

Zinc Coarse Powder, For Analysis, Particle size about 0.3- 1.5mm, EMSURE™, MilliporeSigma™

Catalog No. M1087561000
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1087561000 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1087561000 Supplier Technidata Supplier No. 1.08756.1000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Appearance Silver granules
Quantity 1 kg
Odor Odorless
Packaging Metal can
Form Granular
Formula Weight 65.39
Chemical Name or Material Zinc