accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
EN
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
Sign In
Don't have an account?
Create an Account
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Chemicals
Salts and Inorganics
Inorganic Salts
Post-Transition Metal Salts
ICN225839
Structure Search
Print
Zinc Chromate, MP Biomedicals
Catalog No.
ICN225839
Change view
Chip view icon
List view icon
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
Quantity
ICN225839
100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No.
ICN225839
Supplier
MP Biomedicals Inc
Supplier No.
0522583980
Sign In
or
Register
to check your price and availability.
$146.74
/
Each
Add to Cart
Add to Cart
Description
Specifications
Product Suggestions
Videos
Documents
Certificates
Safety and Handling
Safety Services
Promotions
Specifications
Chemical Identifiers
CAS
13530-65-9
Molecular Formula
CrO4Zn
Molecular Weight (g/mol)
181.37
MDL Number
MFCD00036281
InChI Key
NDKWCCLKSWNDBG-UHFFFAOYSA-N
Synonym
zinc chromate, zinc chrome, zinc chromate c, zinc chromate o, zinc chromate t, zinc chromate z, zinc chromate am, zincro zto, basic zinc chromate, zinc chrome yellow
PubChem CID
26089
IUPAC Name
zinc(2+) dioxochromiumbis(olate)
SMILES
[Zn++].[O-][Cr]([O-])(=O)=O
Specifications
Quantity
100 g
Chemical Name or Material
Zinc Chromate
Product Suggestions
Documents