missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc chloride, 2M solution in 2-MeTHF, AcroSeal™
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Zinc chloride |
| Name Note | 2M solution in 2-Methyltetrahydrofuran |
| Color | Colorless |
| CAS | 7646-85-7, 96-47-9 |
| Physical Form | Liquid |
| Quantity | 100 mL |
| Flash Point | −11°C |
| Packaging | AcroSeal™ Glass Bottle |
| Specific Gravity | 1.07 |
| Density | 1.0700g/mL |
| Show More |
RUO – Research Use Only