missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Zinc-Cast Slotted Masses Set

Alternate for expensive brass weights.

Supplier:  Eisco™ PHZNW500SET

 View more versions of this product

Catalog No. S24450


Only null left

Description

Description

  • Zinc die-casted with powder coat
  • Set of nine weights and one hanger
  • Accuracy ±2%
Specifications

Specifications

Powder-coated Slotted Mass Set
Zinc Die-cast
500 g
Set of 9 x 50g weights and 1 x 50g hanger
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions