missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc carbonate basic, 97%, Zn >58.0%
Description
Zinc carbonate basic is used as pharmaceutical intermediate and in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Zinc carbonate basic is used as pharmaceutical intermediate and in chemical research.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Zinc carbonate basic is used as pharmaceutical intermediate and in chemical research.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Quantity | 1000 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 548.97 |
| Percent Purity | 97% |
| Assay | Zn >58.0% |
| Chemical Name or Material | Zinc carbonate basic |
Safety and Handling
| Hazard Category | H315-H319 |
| Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
| DOTInformation | Hazard Class: 9; Packaging Group: III |
| EINECSNumber | 226-076-7 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only