missing translation for 'onlineSavingsMsg'
Learn More

Zinc Bromide 98.0+%, TCI America™

Catalog No. p-7137992
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
300 g
Catalog No. Quantity
Z001325G 25 g
Z0013300G 300 g
2 options

Catalog No. Z001325G

Supplier: TCI America Z001325G

Only null left

Chemical Identifiers

CAS 7699-45-8
Molecular Formula Br2Zn
Molecular Weight (g/mol) 225.188
MDL Number MFCD00011294
InChI Key VNDYJBBGRKZCSX-UHFFFAOYSA-L
Synonym zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100g
PubChem CID 10421208
IUPAC Name zinc;dibromide
SMILES [Zn+2].[Br-].[Br-]

Specifications

Boiling Point 697°C
Melting Point 394°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
UN Number 3260
Formula Weight 225.19
Percent Purity ≥98.0% (T)
Chemical Name or Material Zinc Bromide
EINECSNumber (1)-0118
RTECSNumber ZH1150000
TSCA Yes