missing translation for 'onlineSavingsMsg'
Learn More

Zinc Bromide Reagent, Reagents

Catalog No. 01336869
Change view
Click to view available options
Quantity: 50 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01336869 Quantity: 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336869 Supplier Labchem Inc Supplier No. C235150050F4

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Zinc Bromide Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7699-45-8
Molecular Formula Br2Zn
Molecular Weight (g/mol) 225.19
InChI Key VNDYJBBGRKZCSX-UHFFFAOYSA-L
IUPAC Name zinc(2+) dibromide
SMILES [Zn++].[Br-].[Br-]

Specifications

Boiling Point 650°C
Color White
pH 5.25
Physical Form Solid
Quantity 50 kg
Grade Reagent
Packaging Fiber Drum
Specific Gravity 4.201
Chemical Name or Material Zinc Bromide Reagent