missing translation for 'onlineSavingsMsg'
Learn More

Zinc bromide, Puriss., anhydrous, ≥98%, Honeywell Fluka™

Catalog No. p-7071697
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
4 x 5 kg
5 kg
Catalog No. Quantity
6001773 1 kg
6001775 5 kg
60044469 4 x 5 kg
3 options

Catalog No. 6001773

Supplier: Honeywell Chemicals 021281KG

Only null left

Puriss., anhydrous,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7699-45-8
Molecular Formula Br2Zn
Molecular Weight (g/mol) 225.188
MDL Number MFCD00011294
InChI Key VNDYJBBGRKZCSX-UHFFFAOYSA-L
Synonym zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100g
PubChem CID 10421208
IUPAC Name zinc;dibromide
SMILES [Zn+2].[Br-].[Br-]

Specifications

Physical Form Anhydrous
Quantity 1 kg
Linear Formula ZnBr2
UN Number 3260
Formula Weight 225.2g/mol
Percent Purity ≥98%
Grade puriss.
Chemical Name or Material Zinc Bromide
Hazard Statement P273-P280-P305 + P351 + P338-P310
Precautionary Statement H302-H314-H317-H411
EINECSNumber 231-718-4