Learn More
Zinc bromide, 99.9% (metals basis)
CAS: 7699-45-8 | Br2Zn | 225.188 g/mol
Supplier: Thermo Scientific Chemicals 04041330
Description
In organic chemistry as a Lewis acid. It is the electrolyte in the zinc bromide battery. Zinc bromide solutions can be used as a transparent shield against radiation. In oil and natural gas wells, solutions containing zinc bromide are used to displace drilling mud.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 650°C | |
| 394°C | |
| Cream to White | |
| 250 g | |
| Br2Zn | |
| UN3260 | |
| zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100g | |
| VNDYJBBGRKZCSX-UHFFFAOYSA-L | |
| zinc;dibromide | |
| 10421208 | |
| 99.9% | |
| Hygroscopic |
| Zinc bromide | |
| 7699-45-8 | |
| Crystalline | |
| (metals basis) | |
| MFCD00011294 | |
| 14,10129 | |
| Very soluble in water,alcohol,ether,acetone,tetrahydrofuran. | |
| [Zn+2].[Br-].[Br-] | |
| 225.188 | |
| 225.19 | |
| 1.5452 | |
| 4.201 g/mL |
Chemical Identifiers
| 7699-45-8 | |
| 225.188 | |
| VNDYJBBGRKZCSX-UHFFFAOYSA-L | |
| 10421208 | |
| [Zn+2].[Br-].[Br-] |
| Br2Zn | |
| MFCD00011294 | |
| zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100g | |
| zinc;dibromide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H302-H314-H317
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 231-718-4
RTECSNumber : ZH1150000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only