missing translation for 'onlineSavingsMsg'
Learn More

Zinc, Activated R, Ricca Chemical

Catalog No. 9432100
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9432100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 9432100 Supplier Ricca Chemical Company Supplier No. R9432000100A6
Only null left

Chemical Identifiers

CAS 7440-66-6 , 16941-12-1
Molecular Weight (g/mol) 65.38 g/mol

Specifications

Quantity 100 g
Packaging White Poly Wide Mouth Bottle
Form Pellet
Grade Laboratory
Chemical Name or Material Zinc, Activated R