missing translation for 'onlineSavingsMsg'
Learn More

Zinc acrylate

Catalog No. AA41780A1
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41780A1 1 kg
AA4178018 50 g
AA4178030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA41780A1 Supplier Thermo Scientific Chemicals Supplier No. 041780A1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14643-87-9 | C6H6O4Zn | 207.49 g/mol

The primary use of Zinc Acrylate is in the production of golf balls. Other uses are primarily in the production of specialty rubber products such as belts and hoses for automobiles. Also used as a tissue adhesive used for adherence of tissue to tissue or tissue to non-tissue for prostheses in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14643-87-9
Molecular Formula C6H6O4Zn
Molecular Weight (g/mol) 207.49
MDL Number MFCD00067208
InChI Key XKMZOFXGLBYJLS-UHFFFAOYSA-L
Synonym zinc acrylate, zinc diacrylate, 2-propenoic acid, zinc salt, acrylic acid, zinc salt, 2-propenoic acid, zinc salt 2:1, zinc prop-2-enoate, 79-10-7 parent, acmc-20aj8t, dsstox_cid_7768, dsstox_rid_78559
PubChem CID 159747
IUPAC Name zinc;prop-2-enoate
SMILES [Zn++].[O-]C(=O)C=C.[O-]C(=O)C=C

Specifications

Melting Point ∼250°C
Density 1.6 g/mL
Quantity 1 kg
UN Number UN3077
Solubility Information Soluble completely in acrylic acid and water (as a salt). Insoluble as a polymer.
Formula Weight 207.5
Physical Form Powder
Chemical Name or Material Zinc acrylate
Hazard Category H302-H314-H317-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 238-692-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only