missing translation for 'onlineSavingsMsg'
Learn More

Zinc acetate dihydrate, Puriss., meets analytical spec. of DAB, E 650, 99-102%, Honeywell Fluka™

Catalog No. 6001770
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001770 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6001770 Supplier Honeywell Chemicals Supplier No. 250441KG
Only null left

Puriss., meets analytical specification of DAB, E 650, 99-102%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 5970-45-6
Molecular Formula C4H10O6Zn
Molecular Weight (g/mol) 219.498
MDL Number MFCD00066961
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
PubChem CID 2724192
IUPAC Name zinc;diacetate;dihydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]

Specifications

Quantity 1 kg
Linear Formula Zn(CH3COO)2 · 2H2O
UN Number 3077
Beilstein 3732513
Formula Weight 219.51g/mol
Percent Purity 99 to 102%
Grade puriss.
Chemical Name or Material Zinc Acetate Dihydrate
Hazard Statement P273-P305 + P351 + P338-P501
Precautionary Statement H302-H319-H410
EINECSNumber 209-170-2