missing translation for 'onlineSavingsMsg'
Learn More

Zinc Acetate, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02004588
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02004588 500 g Poly Bottle
02004587 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02004588 Supplier Avantor Supplier No. 429601

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 5970-45-6
Molecular Formula C4H10O6Zn
Molecular Weight (g/mol) 219.498
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
PubChem CID 2724192
IUPAC Name zinc;diacetate;dihydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]

Specifications

Physical Form Crystals
Quantity 500 g
Packaging Poly Bottle
Specific Gravity 1L = 1.74kg
Formula Weight 219.51
Percent Purity 99.0 to 101.0%
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Zinc Acetate, Dihydrate