missing translation for 'onlineSavingsMsg'
Learn More

Zinc Acetate, Dihydrate, ACS Grade, 98.0% to 101.0%, LabChem™

Catalog No. LC270701
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC270701 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC270701 Supplier LabChem Supplier No. LC270701

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5970-45-6
Molecular Formula C4H10O6Zn
Molecular Weight (g/mol) 219.498
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
PubChem CID 2724192
IUPAC Name zinc;diacetate;dihydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]

Specifications

Melting Point 237°C
Color White
CAS Max % 100
Physical Form Crystals
Quantity 500 g
Linear Formula Zn(C2H3O2)2·2H2O
UN Number UN3077
Solubility Information Soluble in water
Formula Weight 219.49
Percent Purity 98.0 to 101.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1735kg/m3
Decomposition Information Carbon dioxide; Carbon monoxide
Density 1,735kg/m3
Chemical Name or Material Zinc Acetate, Dihydrate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature