missing translation for 'onlineSavingsMsg'
Learn More

Zinc acetate dihydrate, ACS, 98.0-101.0%

Catalog No. p-4782234
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1155930 250 g
AA11559A1 1 kg
AA11559A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1155930 Supplier Thermo Scientific Chemicals Supplier No. 01155930
Only null left

CAS: 5970-45-6 | C4H10O6Zn | 219.498 g/mol

Zinc acetate dihydrate is used in the preparation of layered Zn-arylphosphonates with potential application in sorption, ion exchange or catalysis and zinc sulfide nanoparticles coated on silica particles. It is used in dietary and medical applications. It is also used as lozenges for treating the common cold. It is used as a seeding material to grow zinc oxide nanorods on the microfibers of polyethylene terephthalate fabric. In addition, it finds application as wood preservation, production of other zinc salts, ethylene acetate manufacture, and an analytical reagent in various industries. As a plating inhibitor, it is used in nuclear power plant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5970-45-6
Molecular Formula C4H10O6Zn
Molecular Weight (g/mol) 219.498
MDL Number MFCD00066961
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
PubChem CID 2724192
IUPAC Name zinc;diacetate;dihydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]

Specifications

Melting Point 237°C
Physical Form Crystalline
Quantity 250 g
Assay Percent Range 98 to 101.0%
Beilstein 3732513
Solubility Information Soluble in water,alcoholSoluble in water and alcohol.
Formula Weight 219.50 (183.47 Anhydrous)
Grade ACS
Density 1.735
Chemical Name or Material Zinc Acetate Dihydrate
Hazard Category H302-H319
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing. This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
DOTInformation DOT Class: 9; Packaging Group: III
EINECSNumber 209-170-2
RTECSNumber ZG8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only