missing translation for 'onlineSavingsMsg'
Learn More

Zinc acetate dihydrate, 97+%

Catalog No. AAA12909A4
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA12909A4 2.5 kg
AAA1290922 100 g
AAA1290936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA12909A4 Supplier Thermo Scientific Chemicals Supplier No. A12909A4

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5970-45-6 | C4H10O6Zn | 219.498 g/mol

Zinc acetate dihydrate is used in the preparation of layered Zn-arylphosphonates with potential application in sorption, ion exchange or catalysis and zinc sulfide nanoparticles coated on silica particles. It is used in dietary and medical applications. It is also used as lozenges for treating the common cold. It is used as a seeding material to grow zinc oxide nanorods on the microfibers of polyethylene terephthalate fabric. In addition, it finds application as wood preservation, production of other zinc salts, ethylene acetate manufacture, and an analytical reagent in various industries. As a plating inhibitor, it is used in nuclear power plant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water and alcohol.

Notes
Incompatible with zinc salts, alkalies-their carbonates, oxalates, phosphates and sulfides.

Chemical Identifiers

CAS 5970-45-6
Molecular Formula C4H10O6Zn
MDL Number MFCD00066961
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
PubChem CID 2724192
IUPAC Name zinc;diacetate;dihydrate

Specifications

Melting Point 237°C
Density 1.735
pH 6 to 7
Odor Weak, Characteristic
Quantity 2.5 kg
UN Number UN3077
Beilstein 3732513
Merck Index 14,10128
Solubility Information Soluble in water and alcohol.
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]
Molecular Weight (g/mol) 219.498
Formula Weight 219.50 (183.47 Anhydrous)
Percent Purity ≥97%
Chemical Name or Material Zinc acetate dihydrate
Show More Show Less
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 209-170-2
RTECSNumber ZG8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only